W337609
4-Methylthio-4-methyl-2-pentanone
≥98%, FG
Synonym(s):
cassis pentanone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3SC(CH3)2CH2COCH3
CAS Number:
Molecular Weight:
146.25
FEMA Number:
3376
EC Number:
Council of Europe no.:
11551
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.058
NACRES:
NA.21
Organoleptic:
green; metallic; sulfurous
grade:
FG
Halal
Kosher
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Halal
Kosher
reg. compliance
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110
Assay
≥98%
refractive index
n20/D 1.472 (lit.)
bp
78 °C/15 mmHg (lit.)
density
0.964 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
green; metallic; sulfurous
SMILES string
CSC(C)(C)CC(C)=O
InChI
1S/C7H14OS/c1-6(8)5-7(2,3)9-4/h5H2,1-4H3
InChI key
DHANVOSHQILVNQ-UHFFFAOYSA-N
General description
4-Methylthio-4-methyl-2-pentanone is a volatile sulfur-containing flavor compound found in white wines.
Application
- Optimization of ultrasound assisted-emulsification-dispersive liquid-liquid microextraction by experimental design methodologies for the determination of sulfur compounds in wines by gas chromatography-mass spectrometry.: This paper discusses the optimization of a novel extraction technique using 4-Methylthio-4-methyl-2-pentanone for identifying sulfur compounds in wines. The methodology enhances the detection capabilities of gas chromatography-mass spectrometry, offering significant improvements in food safety and quality control (Silva MF et al., 2010).
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Optimization of ultrasound assisted-emulsification-dispersive liquid?liquid microextraction by experimental design methodologies for the determination of sulfur compounds in wines by gas chromatography?mass spectrometry.
Jofre VP, et al.
Analytica Chimica Acta, 683(1), 126-135 (2010)
Precursors of sulfur?containing flavor compounds
Schutte L & Teranishi R
Critical Reviews in Food Science and Nutrition, 4(4), 457-505 (1974)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service