Skip to Content
Merck
All Photos(1)

Key Documents

W268402

Sigma-Aldrich

Methyl phenylcarbinyl acetate

≥98%, FCC, FG

Synonym(s):

α-Methylbenzyl acetate, α-Methylbenzenemethanol 1-acetate, 1-Phenylethyl acetate, Gardenol, Styrallyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(CH3)C6H5
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2684
EC Number:
Council of Europe no.:
573
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.178
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.494 (lit.)

bp

94-95 °C/12 mmHg (lit.)

density

1.028 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; fruity

SMILES string

CC(OC(C)=O)c1ccccc1

InChI

1S/C10H12O2/c1-8(12-9(2)11)10-6-4-3-5-7-10/h3-8H,1-2H3

InChI key

QUMXDOLUJCHOAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

198.5 °F

Flash Point(C)

92.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S512-S515 (2012-03-22)
A toxicologic and dermatologic review of ethyl phenyl carbinyl acetate when used as a fragrance ingredient is presented. Ethyl phenyl carbinyl acetate is a member of the fragrance structural group Aryl Alkyl Alcohol Simple Acid Esters (AAASAE). The AAASAE fragrance
Guzman Torrelo et al.
Chembiochem : a European journal of chemical biology, 16(4), 625-630 (2015-02-17)
In 2009, we reported that the product of the gene SCJ21.16 (XFa0032) from Xylella fastidiosa, a xylem-restricted plant pathogen that causes a range of diseases in several important crops, encodes a protein (XfHNL) with putative hydroxynitrile lyase activity. Sequence analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service