Unfortunately, there are currently no Dewar flasks available that have been tested for specific heat loss.
SYNF110665
Synthware™ Dewar flask, tall form with metal housing
flask capacity 665 mL, tall form with metal housing
Synonym(s):
Synthware F110665
About This Item
Recommended Products
material
aluminum flask
Joint
Joints neck (70)
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F110665
I.D. × depth
7 cm × 19.5 cm
flask capacity
665 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Dear Sir or Madam, I am looking for a Dewar flask with a specific heat loss per unit mass L (mW/K·kg), which should be around 30. Could you please confirm if the selected product meets this requirement? Thank you in advance for your assistance.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service