Skip to Content
Merck
All Photos(1)

Key Documents

QBD10801

Sigma-Aldrich

m-dPEG®12-Lipoamide

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H65NO13S2
CAS Number:
Molecular Weight:
748.00
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: gold reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCCCC1CCSS1)=O

InChI

1S/C33H65NO13S2/c1-36-9-10-38-13-14-40-17-18-42-21-22-44-25-26-46-29-30-47-28-27-45-24-23-43-20-19-41-16-15-39-12-11-37-8-7-34-33(35)5-3-2-4-32-6-31-48-49-32/h32H,2-31H2,1H3,(H,34,35)

InChI key

WHTUJGAYCNMFSV-UHFFFAOYSA-N

Features and Benefits

The m-dPEG®12-lipoamide has a lipoic acid group linked to a single molecular weight methoxy-terminated dPEG® spacer arm. The lipoic acid group readily forms stable dative bonds with metals such as gold. The hydrophilic, non-immunogenic single molecular weight dPEG® imparts water solubility to the target molecules. As surface modification reagents, these dPEG® products reduce non-specific binding to modified surfaces.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service