GF99049203
Tungsten
wire, 0.457 mm diameter, size 100 mm, condition straight wire
Synonym(s):
W 005457, W
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
wire
manufacturer/tradename
Goodfellow 990-492-03
resistivity
4.9 μΩ-cm, 20°C
size
100 mm
diameter
0.457 mm
bp
5660 °C (lit.)
mp
3410 °C (lit.)
density
19.3 g/mL at 25 °C (lit.)
SMILES string
[W]
InChI
1S/W
InChI key
WFKWXMTUELFFGS-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Experimental neurology, 317, 214-225 (2019-03-12)
Following neurotoxic damage, cells repair their DNA, and survive or undergo apoptosis. This study tests the hypothesis that ethanol induces a DNA damage response (DDR) in neural stem cells (NSCs) that promotes excision repair (ER) and this repair is influenced
Journal of bacteriology, 201(4) (2018-11-21)
At a late stage in spore development in Bacillus subtilis, the mother cell directs synthesis of a layer of peptidoglycan known as the cortex between the two forespore membranes, as well as the assembly of a protective protein coat at
Environmental microbiology, 22(8), 3248-3265 (2020-05-16)
The bacterium Streptomyces davaonensis synthesizes the antibiotic roseoflavin in the stationary phase of growth. The starting point for roseoflavin biosynthesis is riboflavin (vitamin B2 ) and four enzymes (RibCF, RosB, RosA and RosC) are necessary to convert a vitamin (riboflavin)
Molecular cell, 76(6), 953-964 (2019-10-06)
Dynamic protein phosphorylation constitutes a fundamental regulatory mechanism in all organisms. Phosphoprotein phosphatase 4 (PP4) is a conserved and essential nuclear serine and threonine phosphatase. Despite the importance of PP4, general principles of substrate selection are unknown, hampering the study
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service