GF89332780
Stainless Steel - 17-7PH alloy, FeCr17Ni7
foil, 50mm disks, 0.075mm thickness, annealed - condition a, precipitation hardenable
Synonym(s):
FE280300
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11171600
NACRES:
NA.23
form
annealed (condition a: precipitation hardenable)
foil (disks)
manufacturer/tradename
Goodfellow 893-327-80
size × thickness
50 mm × 0.075 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service