Skip to Content
Merck
All Photos(2)

Key Documents

GF40930770

Samarium

lump, 25 mm max. lump size, weight 10 g, purity 95%

Synonym(s):

SM006100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sm
CAS Number:
Molecular Weight:
150.36
MDL number:
UNSPSC Code:
12141612
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

lumps

manufacturer/tradename

Goodfellow 409-307-70

resistivity

91.4 μΩ-cm, 0°C

weight

10 g

max. lump size

25 mm

bp

1794 °C (lit.)

mp

1074 °C (lit.)

density

7.47 g/mL at 25 °C (lit.)

SMILES string

[Sm]

InChI

1S/Sm

InChI key

KZUNJOHGWZRPMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service