GF39538255
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7
1000mm tube, 6.35mm outside diameter, 1.63mm wall thickness, 3.09mm inside diameter, solution treated
Synonym(s):
AL227100
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
tubes
manufacturer/tradename
Goodfellow 395-382-55
L × wall thickness
1000 mm × 1.63 mm
O.D. × I.D.
6.35 mm × 3.09 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service