Skip to Content
Merck
All Photos(1)

Key Documents

CLS707101

Pyrex® VISTA volumetric pipette, Class A

volume 1 mL

Synonym(s):

Corning® 70710

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.43

material

glass

feature

class A
graduation

packaging

case of 12 ea
pack of 12 ea

manufacturer/tradename

Corning 70710-1

volume measuring range

1 mL

capacity

1 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

Pyrex® VISTA volumetric pipettes, Class A (volume 1 mL) are manufactured to Class A capacity tolerances as indicated by ASTM E-969.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service