Skip to Content
Merck
All Photos(2)

Documents

C68105

Sigma-Aldrich

1-Chloro-3-phenylpropane

99%

Synonym(s):

(3-Chloro-1-propyl)benzene, 3-Phenylpropyl chloride, Hydrocinnamyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)3Cl
CAS Number:
Molecular Weight:
154.64
Beilstein:
507635
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.521 (lit.)

bp

219 °C (lit.)

density

1.08 g/mL at 25 °C (lit.)

SMILES string

ClCCCc1ccccc1

InChI

1S/C9H11Cl/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2

InChI key

XZBXAYCCBFTQHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xinguang Zhou et al.
Journal of chromatography. A, 1409, 125-131 (2015-07-25)
Concentration of blood catecholamines (CAs) is linked to a host of cardiovascular diseases, including hypertension and stenocardia. The matrix interferences and low concentration require tedious sample pretreatment methods before quantitative analysis by the gold standard method of high-performance liquid chromatography-electrochemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service