C120006
Cyclopropyl methyl ketone
99%
Synonym(s):
Acetylcyclopropane
About This Item
Recommended Products
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.424 (lit.)
bp
114 °C (lit.)
density
0.849 g/mL at 25 °C (lit.)
SMILES string
CC(=O)C1CC1
InChI
1S/C5H8O/c1-4(6)5-2-3-5/h5H,2-3H2,1H3
InChI key
HVCFCNAITDHQFX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Synthesis of 1,2,4-trioxepanes and 1,2,4-trioxocanes via photooxygenation of homoallylic alcohols.: This study explores the synthesis of novel 1,2,4-trioxepanes and 1,2,4-trioxocanes through the photooxygenation of homoallylic alcohols, utilizing Cyclopropyl methyl ketone as a key reagent in the process. The findings highlight the efficiency and potential applications of this synthetic pathway in organic chemistry and pharmaceutical intermediate production (Singh et al., 2006).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1C
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
69.8 °F - closed cup
Flash Point(C)
21 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service