Key Documents
BR43238
BRAND® density bottle, Gay-Lussac pattern, uncalibrated
capacity 100 mL, borosilicate glass
Synonym(s):
Gay-Lussac
About This Item
UNSPSC Code:
41121800
NACRES:
NB.71
material
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Joint
joint ST/NS neck10/19
feature
with cap
packaging
pack of 2 ea
manufacturer/tradename
BRAND 43238
capacity
100 mL
General description
DIN ISO 3507, type Gay-Lussac. Stopper NS 10/19 with capillary. Top of stopper ground and polished. Nominal capacity printed on the bottom.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.