BR36404
BRAND® BLAUBRAND® volumetric flask trapezoidal
capacity 10 mL, polypropylene stopper, joint: ST/NS 10/19
Synonym(s):
BLAUBRAND®, Volumetric flasks
All Photos(2)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
polypropylene stopper
transparent borosilicate glass 3.3 flask
Joint
joint: ST/NS10/19
feature
accuracy class A
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36404
capacity
10 mL
General description
Borosilicate 3.3. To DIN EN ISO 1042. Calibrated to contain (TC, In).
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG