BAF369100060
SP Bel-Art® Sterileware™ Pharma Scoop
red polystyrene, nominal capacity 60 mL (2 oz), sterile, pack of 100 ea (individually wrapped)
Synonym(s):
Lab Scoop, spoon
All Photos(1)
About This Item
Recommended Products
material
red polystyrene
sterility
sterile
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
Bel-Art F36910-0060
bowl H × L × W
1.125 in. (28 mm) × 2.875 in. (72 mm) × 2 in. (49 mm)
nominal capacity
60 mL (2 oz)
General description
Ideal for sampling for both production work and laboratory use, Pharma Scoops are used by pharmaceutical, food, cosmetic, and chemical companies.
Features and Benefits
- High handle location keeps hand further from sample
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)Ready for one time use and disposal
- Lot stamped for accurate tracing
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP