Skip to Content
Merck
All Photos(1)

Documents

935042

Sigma-Aldrich

AC1 Screen Printed Electrodes

20 units (2mm Gold Working; Gold Counter; Silver Reference)

Synonym(s):

SP-1112

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NA.21

General description

The sensor is formed on a corundum ceramic base. On to this surface the working, the reference and the auxiliary electrodes are applied. The working and the auxiliary electrodes are made of variety of materials. At the end of the sensor there is a contacting field which is connected with the active part by the silver conducting paths which are covered by a dielectric protection layer. A biochemically active substance can be immobilised on the working electrode of the sensor to create a biosensor.
We offer Screen Printed Electrodes (2mm Gold Working; Gold Counter; Silver Reference) for R&D applications. The physical parameters are as follows:
Dimensions:Weight: 0.5 gm; Length: 25.40 mm; Width: 7.26 mm; Thickness: 0.63 mm

Application

Screen printed electrodes (2mm Gold Working; Gold Counter; Silver Reference) are ideal for electrochemical detection. The various application areas of Screen printed electrodes (SPEs) are: (1) for the detection of common environmental pollutants such as metal ions and organic compounds; (2) as enzymatic biosensors, immunosensors and DNA-based sensors for pharmaceutical and clinical usage; (3) also in emerging applications of fabricating lithium-ion batteries, solar cells, and large area LED matrix.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screen printing of stretchable electrodes for large area LED matrix
Ho Xinning, et al.
Journal of Materials Research, 30, 2271?2278-2271?2278 (2015)
Applications of screen-printed electrodes in current environmental analysis
Li M, et al.
Current Opinion in Electrochemistry, 3, 137-143 (2017)
High performance screen-printed electrodes prepared by a green solvent approach for lithium-ion batteries
Goren A., et al.
Journal of Power Sources, 334, 65-77 (2016)
Screen printed silver top electrode for efficient inverted organic solar cells
Kim Junwoo, et al.
Materials Research Bulletin, 70, 412-415 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service