Skip to Content
Merck
All Photos(2)

Documents

932671

Sigma-Aldrich

4,7-Diphenyl-1,10-phenanthroline

Synonym(s):

Bathophenanthroline, 4,7-Diphenyl-1,10-phenanthroline, BPhen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N2
CAS Number:
Molecular Weight:
332.40
Beilstein:
261048
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.21

grade

sublimed grade

Quality Level

description

µe ≈ 3.0 x 10-4 cm2V-1s-1

Assay

≥99% (H-NMR)

form

powder

loss

0.5% TGA, >240ºC (weight loss)

mp

218-220 °C (lit.)

solubility

THF: soluble
chloroform: soluble
dichloromethane: soluble

λmax

272 nm in THF

fluorescence

λem 379 nm in THF

Orbital energy

HOMO 6.4 eV 
LUMO 3.0 eV 

SMILES string

c1ccc(cc1)-c2ccnc3c2ccc4c(ccnc34)-c5ccccc5

InChI

1S/C24H16N2/c1-3-7-17(8-4-1)19-13-15-25-23-21(19)11-12-22-20(14-16-26-24(22)23)18-9-5-2-6-10-18/h1-16H

InChI key

DHDHJYNTEFLIHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intermolecular charge-transfer states at the interface between electron donating (D) and accepting (A) materials are crucial for the operation of organic electronics, such as solar cells, transistors and organic light-emitting diodes (OLEDs). The 4,7-Diphenyl-1,10-phenanthroline, also known as Bathophenanthroline (Bphen) enables use as Electron Transport / Hole Blocking Layer (ETL / HBL) in your organic electronic devices. It has a μe of about 3.0 x 10-4 cm2 V−1 s−1 and is solution-processable.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strong light-matter coupling for reduced photon energy losses in organic photovoltaics
Nikolis, V. et al.
Nature Communications, 10, 1-8 (2019)
Highly efficient inverted organic light-emitting devices adopting solution-processed double electron-injection layers.
Chen, Y. et al.
Organic Electronics, 66, 1-6 (2019)
Light outcoupling efficiency enhancement in organic light emitting diodes using an organic scattering layer
Grover Rakhi, et al.
Physica Status Solidi RRL: Rapid Research Letters, 8, 81?85-81?85 (2014)
Emissive and charge-generating donor-acceptor interfaces for organic optoelectronics with low voltage losses
Ullbrich, S. et al.
Nature Materials, 18, 459-464 (2019)
Initial photochemical stability in perovskite solar cells based on the Cu electrode and the appropriate charge transport layers
Tan, Wenjun, et al.
Synthetic Metals, 246, 101-107 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service