Skip to Content
Merck
All Photos(1)

Key Documents

909890

Sigma-Aldrich

[1,1′:4′,1″-Terphenyl]-3,3″- dicarboxylic acid, 2,2″,4,4″,6,6″- hexamethyl-, 3,3″-dimethyl ester

≥97%

Synonym(s):

HMTE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30O4
CAS Number:
Molecular Weight:
430.54
UNSPSC Code:
43211915
NACRES:
NA.23

Assay

≥97% (HPLC)
≥97%

form

powder

color

white to off-white

storage temp.

2-8°C

Application

HMTE is a diester monomer used for developing high-performance polyesters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service