Skip to Content
Merck
All Photos(3)

Key Documents

905321

Sigma-Aldrich

Rh2(R-TCPTAD)4

Synonym(s):

Davies dirhodium catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H64Cl16N4O16Rh2
CAS Number:
Molecular Weight:
2110.44
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

form

powder, crystals or chunks

Application

This dirhodium catalyst developed by the Davies lab can form C-C bonds at the most accessible tertiary C-H position with control of both regioselectivity and absolute configuration.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuangbiao Liao et al.
Nature, 551(7682), 609-613 (2017-11-21)
The synthesis of complex organic compounds usually relies on controlling the reactions of the functional groups. In recent years, it has become possible to carry out reactions directly on the C-H bonds, previously considered to be unreactive. One of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service