Skip to Content
Merck
All Photos(2)

Key Documents

904449

Sigma-Aldrich

Silica

monodisperse, non-porous, 500 nm

Synonym(s):

Monodisperse silicon dioxide, Non-porous silica, Non-porous silicon dioxide, Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
41123003
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monodisperse nonporous silica nanoparticles find application in drug delivery and molecular imaging[1][2] whereas micron sized particles are used in liquid chromatography.[3] We offer highly monodisperse(coefficient of variance <10%) nonporous silica in size varying from 150 nm to 50 μm. The high monodispersity of our particles makes them suitable for aforementioned applications.
Silica monodispersed, non-porous (500 nm) is a versatile filler material that can be used in a variety of applications such as catalysts, coatings, and absorbents.[4] It can also be used in the development of nanomedicine for biomedical and clinical applications.[5][6]
Silica monodispersed, non-porous (550 nm) is a versatile filler material that can be used in a variety of applications such as catalysts, coatings, and absorbents.[4] It can also be used in the development of nanomedicine for biomedical and clinical applications.[5][6]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical Examination of a Nonporous Silica Stationary Phase for Reversed-Phase Fast LC Applications
D R Jenke
Journal of Chromatographic Science, 34, 362- 362 (1999)
Thermal pretreatment of silica composite filler materials
Wan Q, et al.
Journal of Thermal Analysis and Calorimetry, 99(1), 237-243 (2009)
The effects of nonporous silica nanoparticles on cultured human keratocytes
Yim B, et al.
Investigative Ophthalmology & Visual Science, 58(1), 362-371 (2017)
Nonporous silica nanoparticles for nanomedicine application
Tang L and Cheng J
Nano Today, 8(3), 290-312 (2013)
Martha Kim et al.
Scientific reports, 7(1), 8238-8238 (2017-08-16)
Nonporous silica nanoparticles (SiNPs) have potential as promising carriers for ophthalmic drugs. However, the in vivo safety of ocular topical SiNPs remains unclear. This study investigated the in vivo safety of oral and ocular topical applications of 100 nm-sized SiNPs in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service