901397
Poly(ethylene glycol) linear dendrimer
NHBoc terminated, PEG Mn 6000, generation 4
Synonym(s):
Dendron bifunctionalized PEG, PFD-G4-PEG6k-NHBoc, Polyethylene glycol
About This Item
Recommended Products
General description
- Polymer architecture: Dendron-functionalized PEG
- Generation: 4
- PEG Average Mn: 6,000
- End group functionality: Boc-protected amine
- No. surface groups: 32
- Calculated mol. wt.: 14970.98 g/mol
- Boc: tert-Butyloxycarbonyl
- Bis-MPA: 2,2-bis(hydroxymethyl)propionic acid
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).
Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).
Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).
Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service