Skip to Content
Merck
All Photos(1)

Key Documents

806099

Sigma-Aldrich

bis-MPA-COOH dendrimer

trimethylol propane core, generation 1

Synonym(s):

G1-TMP-COOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H62O30
Molecular Weight:
1082.96
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

solid or liquid

storage temp.

−20°C

SMILES string

O=C(OCC(CC)(COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)COC(C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C)=O)C(COC(CCC(O)=O)=O)(COC(CCC(O)=O)=O)C

InChI

1S/C45H62O30/c1-5-45(24-73-39(64)42(2,18-67-33(58)12-6-27(46)47)19-68-34(59)13-7-28(48)49,25-74-40(65)43(3,20-69-35(60)14-8-29(50)51)21-70-36(61)15-9-30(52)53)26-75-41(66)44(4,22-71-37(62)16-10-31(54)55)23-72-38(63)17-11-32(56)57/h5-26H2,1-4H3,(H,46,47)(H,48,49)(H,50,51)(H,52,53)(H,54,55)(H,56,57)

InChI key

PAYWQMZAUNWKEB-UHFFFAOYSA-N

General description

  • End Group Functionality: Carboxyl surface group
  • Generation: 1
  • No. Surface Groups: 6
  • Calculated Mol. Wt.: 1083 g/mol
  • bis-MPA = 2,2-bis(hydroxymethyl)propionic acid

Application

Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irena Mamajanov et al.
Life (Basel, Switzerland), 10(8) (2020-08-23)
Enzymes are biological catalysts that are comprised of small-molecule, metal, or cluster catalysts augmented by biopolymeric scaffolds. It is conceivable that early in chemical evolution, ancestral enzymes opted for simpler, easier to assemble scaffolds. Herein, we describe such possible protoenzymes:
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service