Skip to Content
Merck
All Photos(1)

Key Documents

771058

Sigma-Aldrich

4-(4′-Hydroxyphenylazo)benzoic acid

97%

Synonym(s):

(E)-4-[(4-Hydroxyphenyl)azo]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O3
CAS Number:
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

270-280 °C

functional group

carboxylic acid
hydroxyl

storage temp.

2-8°C

SMILES string

OC(C1=CC=C(/N=N/C2=CC=C(O)C=C2)C=C1)=O

InChI

1S/C13H10N2O3/c16-12-7-5-11(6-8-12)15-14-10-3-1-9(2-4-10)13(17)18/h1-8,16H,(H,17,18)/b15-14+

InChI key

HLVCZTOFOWHIJZ-CCEZHUSRSA-N

Application

4-(4′-Hydroxyphenylazo)benzoic acid may be used to synthesize 4-(4-propyloxyphenylazo)benzoic acid via Williamson synthesis by reacting with 1-bromopropane.
Azobenzene cleavable linker. Treatment with sodium dithionite (sodium hydrosulfite) reduces azo functionality, cleaving the N-N bond to yield two primary amines.
Has been used in proteomic and affinity chromatography applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular imprinting of azobenzene carboxylic acid on a TiO2 ultrathin film by the surface sol-gel process
Lee SW, et al.
Langmuir, 14(10), 2857-2863 (1998)
A mild chemically cleavable linker system for functional proteomic applications.
Steven H L Verhelst et al.
Angewandte Chemie (International ed. in English), 46(8), 1284-1286 (2007-01-06)
Felicetta Landi et al.
Organic & biomolecular chemistry, 8(1), 56-59 (2009-12-22)
A new chemically-cleavable linker has been synthesised for the affinity-independent elution of biomolecules by classical affinity chromatography. This azo-based linker is shown to couple efficiently with "click" derivatised ligands such as biotin propargyl amide through a copper(I)-catalysed Huisgen 1,3-dipolar cycloaddition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service