Skip to Content
Merck
All Photos(2)

Key Documents

768995

Sigma-Aldrich

Helium

Messer® CANGas, 99.999%

Synonym(s):

Helium gas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
He
CAS Number:
Molecular Weight:
4.00
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

0.14 (vs air)

Quality Level

description

Filling mass: 2g
Filling volume: 12L

Assay

99.999%

form

gas

bp

-268.934 °C (lit.)
−268.9 °C (lit.)

mp

-272.2 °C (lit.)

SMILES string

[He]

InChI

1S/He

InChI key

SWQJXJOGLNCZEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Helium is a colourless and odourless rare gas that is much lighter than air. Application areas include leak detection, aeronautics, metallurgy, laser welding and technology, and meteorology (weather balloons). Helium is also used as a shielding gas, reactor cooling gas, carrier gas for gas chromatography, and cooling gas for production of optical fibres. It is also a filling gas for air bags and is a component of diving mixtures. Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service