Skip to Content
Merck
All Photos(1)

Key Documents

711004

Sigma-Aldrich

Resveratrol-(4-hydroxyphenyl-13C6)

99 atom % 13C, 98% (CP)

Synonym(s):

3,4′,5-Trihydroxy-trans-stilbene-13C6, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C8H12O3
CAS Number:
Molecular Weight:
234.20
MDL number:
UNSPSC Code:
12191503
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

Oc1cc(O)cc(\C=C\[13c]2[13cH][13cH][13c](O)[13cH][13cH]2)c1

InChI

1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1+/i3+1,4+1,5+1,6+1,10+1,12+1

InChI key

LUKBXSAWLPMMSZ-OYESDQHJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service