Skip to Content
Merck
All Photos(1)

Documents

68522

Sigma-Aldrich

trans-4-Methyl-2-pentene

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1719115
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.392

density

0.686 g/mL at 20 °C (lit.)

SMILES string

C\C=C\C(C)C

InChI

1S/C6H12/c1-4-5-6(2)3/h4-6H,1-3H3/b5-4+

InChI key

LGAQJENWWYGFSN-SNAWJCMRSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service