Skip to Content
Merck
All Photos(2)

Key Documents

682705

Sigma-Aldrich

Chloro(tri-tert-butylphosphine)gold(I)

Synonym(s):

(Tri-tert-butylphosphine)chlorogold, (Tri-tert-butylphosphine)gold(I) chloride, (tBu3P)AuCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H9)3PAuCl
CAS Number:
Molecular Weight:
434.74
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

>300 °C

SMILES string

Cl[Au].CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/C12H27P.Au.ClH/c1-10(2,3)13(11(4,5)6)12(7,8)9;;/h1-9H3;;1H/q;+1;/p-1

InChI key

JLXSZGSXDDQSJF-UHFFFAOYSA-M

Application

Gold Catalysts — 21st Century ′Gold Rush′

  • Useful catalyst in a variety of C-C bond forming reactions, including cyclopropanation.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toste, F.D. et al.
Journal of the American Chemical Society, 172, 18002-18002 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service