Skip to Content
Merck
All Photos(1)

Documents

656682

Sigma-Aldrich

Chloromethyl chlorosulfate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2OSO2Cl
CAS Number:
Molecular Weight:
165.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.448

bp

45-50 °C/10 mmHg

density

1.631 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

ClCOS(Cl)(=O)=O

InChI

1S/CH2Cl2O3S/c2-1-6-7(3,4)5/h1H2

InChI key

PJBIHXWYDMFGCV-UHFFFAOYSA-N

Application

Reagent for chloromethylation of Fmoc-protected amino acids without the generation of carcinogenic bis(chloromethyl)ether.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Carc. 1A - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nicholas P Power et al.
Organic & biomolecular chemistry, 2(10), 1554-1562 (2004-05-12)
The reaction of liquid (gamma-) SO3 with CH2Cl2 at room temperature leads to SO3 insertion into the C-Cl bonds, giving the useful chloromethylating agent chloromethyl chlorosulfate (CMCS). The process is very slow but becomes rapid on addition of catalytic quantities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service