Skip to Content
Merck
All Photos(1)

Key Documents

631175

Sigma-Aldrich

Methyl 6-chloropyridine-3-carboxylate

98%

Synonym(s):

Methyl 6-chloronicotinate, Methyl 6-chloro-3-picolinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

86-90 °C (lit.)

SMILES string

COC(=O)c1ccc(Cl)nc1

InChI

1S/C7H6ClNO2/c1-11-7(10)5-2-3-6(8)9-4-5/h2-4H,1H3

InChI key

RMEDXVIWDFLGES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 6-chloropyridine-3-carboxylate can be used to synthesize fluoro substituted 6-phenylnicotinamide, which shows TRPV1 antagonist potency. It can also be used in the synthesis of retinoid x receptor (RXR) ligands, which have many clinical applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Replacing alkyl sulfonamide with aromatic sulfonamide in sulfonamide-type RXR agonists favors switch towards antagonist activity"
Morishita I-K, et al.
Bioorganic & Medicinal Chemistry, 19(03), 1001-1003 (2009)
"Design and synthesis of 6-phenylnicotinamide derivatives as antagonists of TRPV1"
Westaway.MS, et al.
Bioorganic & Medicinal Chemistry, 18(20), 5609-5613 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service