Skip to Content
Merck
All Photos(1)

Key Documents

604887

Sigma-Aldrich

L-Serine-13C3

98 atom % 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO13CH213CH(NH2)13CO2H
CAS Number:
Molecular Weight:
108.07
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % 13C

Assay

95% (CP)

form

solid

optical activity

[α]25/D +14.6°, c = 2 in 1 M HCl

mp

222 °C (dec.) (lit.)

mass shift

M+3

SMILES string

N[13C@@H]([13CH2]O)[13C](O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1/i1+1,2+1,3+1

InChI key

MTCFGRXMJLQNBG-GCCOVPGMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service