Skip to Content
Merck
All Photos(1)

Documents

560839

Sigma-Aldrich

3-Vinylaniline

97%

Synonym(s):

3-Aminostyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHC6H4NH2
CAS Number:
Molecular Weight:
119.16
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

contains

KOH as inhibitor

refractive index

n20/D 1.611 (lit.)

density

1.051 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1cccc(C=C)c1

InChI

1S/C8H9N/c1-2-7-4-3-5-8(9)6-7/h2-6H,1,9H2

InChI key

IFSSSYDVRQSDSG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Zarco-Fernández et al.
Talanta, 162, 612-617 (2016-11-14)
A new selective polymer has been developed (Patent: P201500704) with high mercury sorption capacity. The combination of this material with a Direct Mercury Analyser (DMA80) has overcome important limitations of this equipment for mercury determination of complex matrix samples, like
H Al-Sagur et al.
Biosensors & bioelectronics, 92, 638-645 (2016-11-12)
Herein, we report a scalable synthesis of multifunctional conducting polyacrylic acid (PAA) hydrogel (MFH) integrated with reduced grapheme oxide (rGO), vinyl substituted polyaniline (VS-PANI) and lutetium Phthalocyanine (LuPc

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service