Skip to Content
Merck
All Photos(3)

Key Documents

496847

Sigma-Aldrich

Tetrahydrofuran-3-carboxaldehyde solution

50 wt. % in H2O, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

concentration

50 wt. % in H2O

refractive index

n20/D 1.416

bp

99 °C

density

1.106 g/mL at 25 °C

functional group

aldehyde
ether

storage temp.

2-8°C

SMILES string

O=CC1CCOC1

InChI

1S/C5H8O2/c6-3-5-1-2-7-4-5/h3,5H,1-2,4H2

InChI key

GSUBXIVOZXWGKF-UHFFFAOYSA-N

General description

The product is a 50 wt.% solution of tetrahydrofuran-3-carboxaldehyde in water.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service