Skip to Content
Merck
All Photos(1)

Key Documents

476668

Sigma-Aldrich

Bromobis(dimethylamino)borane

97%

Synonym(s):

1-Bromo-N,N,N′,N′-tetramethylboranediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]2BBr
CAS Number:
Molecular Weight:
178.87
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Quality Level

Assay

97%

reaction suitability

reagent type: reductant

refractive index

n20/D 1.478 (lit.)

bp

20-28 °C/0.5 mmHg (lit.)

density

1.259 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)B(Br)N(C)C

InChI

1S/C4H12BBrN2/c1-7(2)5(6)8(3)4/h1-4H3

InChI key

UBDPZBLACZIAPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Intramolecular cyclization
  • Oxidative addition for synthesis of platinum boryl phosphine bromo complexes
  • Synthesis of fluorenyltripyrazolylborate ligands
  • Preparation of rigid-backbone conjugated polymers
  • Conversion of epoxides to halohydrins

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

129.0 °F - closed cup

Flash Point(C)

53.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service