Skip to Content
Merck
All Photos(1)

Key Documents

464929

Sigma-Aldrich

5-Hexenoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CH(CH2)3CO2H
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.435 (lit.)

bp

105 °C/15 mmHg (lit.)

density

0.961 g/mL at 25 °C (lit.)

functional group

allyl
carboxylic acid

SMILES string

OC(=O)CCCC=C

InChI

1S/C6H10O2/c1-2-3-4-5-6(7)8/h2H,1,3-5H2,(H,7,8)

InChI key

XUDOZULIAWNMIU-UHFFFAOYSA-N

Application

5-Hexenoic acid may be used for the preparation of iodolactone derivatives and fluorine-containing δ-valerolactones.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tertiary aminourea-catalyzed enantioselective iodolactonization.
Gemma E Veitch et al.
Angewandte Chemie (International ed. in English), 49(40), 7332-7335 (2010-08-31)
Stilianos G Roussis et al.
Journal of chromatography. A, 1594, 105-111 (2019-03-02)
In an effort to improve separation of impurities in oligonucleotide drugs, alkyl amines of different length and carbon content were evaluated as reagents in ion pair-reversed phase (IP-RP) HPLC with mass spectrometric detection. A range of columns was tested in
Synthesis of fluoroalkyl-d-lactones from polyfluoroalkyl iodides and 5-hexenoic acids.
Fang X, et al.
Journal of Fluorine Chemistry, 129(4), 280-285 (2008)
Philipp Höfer et al.
Microbial cell factories, 9, 70-70 (2010-09-18)
Methylotrophic (methanol-utilizing) bacteria offer great potential as cell factories in the production of numerous products from biomass-derived methanol. Bio-methanol is essentially a non-food substrate, an advantage over sugar-utilizing cell factories. Low-value products as well as fine chemicals and advanced materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service