Skip to Content
Merck
All Photos(1)

Documents

394262

Sigma-Aldrich

Octadecylmagnesium chloride solution

0.5 M in THF

Synonym(s):

Chlorooctadecylmagnesium, Octadecane, magnesium complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17MgCl
CAS Number:
Molecular Weight:
313.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

67 °C

density

0.906 g/mL at 25 °C

SMILES string

CCCCCCCCCCCCCCCCCC[Mg]Cl

InChI

1S/C18H37.ClH.Mg/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2;;/h1,3-18H2,2H3;1H;/q;;+1/p-1

InChI key

ONJPXWZQVLKGIU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Octadecylmagnesium chloride is a Grignard reagent commonly used in organic synthesis for various transformations, including nucleophilic addition reactions and carbon-carbon bond formation via palladium-catalyzed alkylation and arylation reactions.

Application

Octadecylmagnesium chloride can be used for the introduction of octyl substituents on the bipyridine skeletons to prepare functionalized 4,4′-bipyridines, which are used as linkers formation of discrete metal complexes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service