Skip to Content
Merck
All Photos(1)

Key Documents

33514

Sigma-Aldrich

Phenol – 1,1,2,2-tetrachloroethane mixture

60:40% (w/w)

Synonym(s):

1,1,2,2-Tetrachloroethane – phenol mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Assay

59.5—60.5% (GC)

form

liquid

composition

phenol, 59.5-60.5 wt. % GC
tetrachloroethane, 39.5-40.5 wt. % GC

color

clear colorless

SMILES string

ClC(Cl)C(Cl)Cl.Oc1ccccc1

InChI

1S/C6H6O.C2H2Cl4/c7-6-4-2-1-3-5-6;3-1(4)2(5)6/h1-5,7H;1-2H

InChI key

SLJYHQQNQRQZDO-UHFFFAOYSA-N

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.
Mixture may demix upon storage

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service