Skip to Content
Merck
All Photos(1)

Key Documents

245860

Sigma-Aldrich

Methyl 4-chloroacetoacetate

97%

Synonym(s):

γ-Chloroacetoacetic acid methyl ester, 4-Chloro-3-oxo-butyric acid methyl ester, 4-Chloro-3-oxobutanoic acid methyl ester, Methyl γ-chloroacetoacetate, Methyl (chloroacetyl)acetate, Methyl 3-oxo-4-chlorobutanoate, Methyl 4-chloro-3-oxobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COCH2COOCH3
CAS Number:
Molecular Weight:
150.56
Beilstein:
1759939
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

85 °C/4 mmHg (lit.)

density

1.305 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CC(=O)CCl

InChI

1S/C5H7ClO3/c1-9-5(8)2-4(7)3-6/h2-3H2,1H3

InChI key

HFLMYYLFSNEOOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reacts with symmetric allylic diacetates or dicarbonates in the presence of a palladium catalyst to give dihyrofurans.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 39-39 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service