Skip to Content
Merck
All Photos(1)

Key Documents

24350

Sigma-Aldrich

2-Chloro-N,N-dimethylacetamide

≥97.0% (GC)

Synonym(s):

N,N-Dimethylchloroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CON(CH3)2
CAS Number:
Molecular Weight:
121.57
Beilstein:
969568
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.479

density

1.182 g/mL at 20 °C (lit.)

functional group

amide
chloro

SMILES string

CN(C)C(=O)CCl

InChI

1S/C4H8ClNO/c1-6(2)4(7)3-5/h3H2,1-2H3

InChI key

XBPPLECAZBTMMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloro-N,N-dimethylacetamide (N,N-dimethylchloroacetamide) was used in the synthesis of (S)-carbinoxamine[1].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of (S)-carbinoxamine. New aspects of oxazaborolidine-catalyzed enantioselective carbonyl reduction.
Corey EJ and Helal CJ.
Tetrahedron Letters, 37(32), 5675-5678 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service