Skip to Content
Merck
All Photos(1)

Key Documents

177822

Sigma-Aldrich

2,2,2-Trifluoroethyl p-toluenesulfonate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3CH2CF3
CAS Number:
Molecular Weight:
254.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

87-92 °C/0.1 mmHg (lit.)

mp

36-38 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCC(F)(F)F

InChI

1S/C9H9F3O3S/c1-7-2-4-8(5-3-7)16(13,14)15-6-9(10,11)12/h2-5H,6H2,1H3

InChI key

IGKCQDUYZULGBM-UHFFFAOYSA-N

Application

2,2,2-Trifluoroethyl p-toluenesulfonate was used in the synthesis of 1-alkyl-2,2-difluorovinylboranes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A one-pot synthesis of 2, 2-difluorovinyl carbonyl compounds from 2, 2, 2,-trifluoroethyl p-toluenesulfonate via 2, 2-difluorovinylboranes.
Tetrahedron Letters, 33(3), 337-340 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service