Skip to Content
Merck
All Photos(1)

Documents

151963

Sigma-Aldrich

Nitromethane-d3

99 atom % D

Synonym(s):

Trideuteronitromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3NO2
CAS Number:
Molecular Weight:
64.06
Beilstein:
1751076
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

expl. lim.

7.1-63 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.1000% water
water

refractive index

n20/D 1.3795 (lit.)

bp

100 °C (lit.)

mp

-29 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])[N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3/i1D3

InChI key

LYGJENNIWJXYER-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitromethane-d3 is a deuterated derivative of nitromethane. Its pyrolytic decomposition in the absence and presence of nitrogen dioxide have been studied. Molecular structure studies suggest that its molecule is composed of a highly asymetrical top consisting of a heavy planar frame of NO2 (C2v symmetry) and a lighter top of CD3 (C3v symmetry). Rotational constants, high-resolution Fourier transform infrared (FTIR) spectrum and DFT (Density Functional Theory) calculations of nitromethane-d3 have been evaluated. Various physicochemical properties (densities, viscosities, speed of sound and refractive indices) of CD3NO2 have been evaluated in the temperature range of 293K to 308K.

Application


  • Asymmetric transformation of a double-stranded, dicopper(I) helicate containing achiral bis(bidentate) Schiff bases.: This study used Nitromethane-d₃ to investigate the asymmetric transformation in dicopper(I) helicates, providing insights into chiral synthesis and coordination chemistry applications (Habermehl et al., 2006).

  • Reversing a rotaxane recognition motif: threading oligoethylene glycol derivatives through a dicationic cyclophane.: The research employed Nitromethane-d₃ in the synthesis and study of rotaxanes, focusing on molecular recognition and self-assembly processes important for developing advanced materials and nanotechnology (Chiu and Stoddart, 2002).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrolysis of nitromethane-d3.
Crawforth CG and Waddington DJ.
The Journal of Physical Chemistry, 74(4), 2793-2796 (1970)
Isotope effects on selected physicochemical properties of nitromethane and 1-pentanol.
Makowska A and Szydlowski J.
Nukleonika, 51, 81-85 (2006)
DFT molecular structure and initial assignment for the FTIR spectrum of ?7 band of nitromethane-D3.
Pal C and Kshirsagar RJ.
Indian J. Phys., 84(8), 929-937 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service