Skip to Content
Merck
All Photos(3)

Documents

136867

Sigma-Aldrich

2,4-Dimethyl-3-pentanone

98%

Synonym(s):

Diisopropyl ketone, Isobutyrone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOCH(CH3)2
CAS Number:
Molecular Weight:
114.19
Beilstein:
773782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.400 (lit.)

bp

124 °C (lit.)

mp

−80 °C (lit.)

density

0.806 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(=O)C(C)C

InChI

1S/C7H14O/c1-5(2)7(8)6(3)4/h5-6H,1-4H3

InChI key

HXVNBWAKAOHACI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dimethyl-3-pentanone was used in an experimental method to measure the equilibrium isotopic fractionation factor (αeq ) for C-bound H positions adjacent to carbonyl group in ketones.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydroboration of acyclic allenes with disiamylborane.
Sethi DS, et al.
Canadian Journal of Chemistry, 47(6), 1083-1086 (1969)
Equilibrium 2H/1H fractionations in organic molecules: I. Experimental calibration of ab initio calculations.
Wang Y, et al.
Geochimica et Cosmochimica Acta, 73(23), 7060-7075 (2009)
Yu Song et al.
The Journal of chemical physics, 142(22), 224306-224306 (2015-06-15)
Ultraviolet (UV) photodissociation dynamics of jet-cooled n-propyl (n-C3H7) radical via the 3s Rydberg state and i-propyl (i-C3H7) radical via the 3p Rydberg states are studied in the photolysis wavelength region of 230-260 nm using high-n Rydberg atom time-of-flight and resonance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service