Skip to Content
Merck
All Photos(2)

Documents

127124

Sigma-Aldrich

N,N,N′-Trimethylethylenediamine

97%

Synonym(s):

1-(Dimethylamino)-2-(methylamino)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2NHCH3
CAS Number:
Molecular Weight:
102.18
Beilstein:
505989
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.419 (lit.)

bp

116-118 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

SMILES string

CNCCN(C)C

InChI

1S/C5H14N2/c1-6-4-5-7(2)3/h6H,4-5H2,1-3H3

InChI key

HVOYZOQVDYHUPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N,N′-Trimethylethylenediamine is synthesized using laser-vaporization supersonic molecular beam. It has been studied using pulsed-field ionization zero electron kinetic energy.

Application

N,N,N′-Trimethylethylenediamine has been used in the preparation of a novel gemini surfactant, N,N-dimethyl-N-{2-[N′-methyl-N′-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides (2C(n)AmSb, where n represents hydrocarbon chain lengths of 8, 10, 12, and 14), by reacting it with n-alkyl bromide. It has also been used as an amine component for in situ formation of α-amino alkoxides.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Wang et al.
The journal of physical chemistry. A, 110(24), 7568-7576 (2006-06-16)
Copper complexes of ethylenediamine (en), N-methylethylenediamine (meen), N,N-dimethylethylenediamine (dmen), N,N,N'-trimethylethylenediamine (tren), and N,N,N',N'-tetramethylethylenediamine (tmen) are synthesized in laser-vaporization supersonic molecular beams and studied by pulsed-field ionization zero electron kinetic energy (ZEKE) and photoionization efficiency spectroscopies and second-order Moller-Plesset perturbation theory.
Santanu Patra et al.
Biomaterials science, 4(3), 418-429 (2015-12-04)
To overcome the problems associated with conventional liposomes in transdermal drug delivery like limited penetration ability and poor stability, in this article we report a new generation of cell penetrating peptide polyarginine containing nano-liposomes conjugated with carbon dots. The newly
Irina-Claudia Grig-Alexa et al.
Organic letters, 8(19), 4187-4189 (2006-09-08)
The synthesis of new 4,6-disubstituted dihydrodipyridopyrazines starting from corresponding carboxaldehydes via lithiation directed by alpha-amino alkoxides is described. The N,N,N'-trimethylethylenediamine was used as amine component for in situ formation of the alpha-amino alkoxides. After optimization, this reaction allowed easy access
Kanae Nyuta et al.
Journal of colloid and interface science, 301(1), 267-273 (2006-05-30)
A novel gemini surfactant with two hydrocarbon chains and two different hydrophilic parts containing a quaternary ammonium salt and a sulfobetaine moiety, N,N-dimethyl-N-{2-[N'-methyl-N'-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides (2C(n)AmSb, where n represents hydrocarbon chain lengths of 8, 10, 12, and 14), was synthesized by
J M Entrena et al.
Scientific reports, 6, 37835-37835 (2016-11-26)
Sigma-1 receptor antagonists promote antinociception in several models of pain, but the effects of sigma-1 agonists on nociception (particularly when the nociceptive system is primed) are not so well characterized; therefore we evaluated the effects of sigma-1 agonists on pain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service