Skip to Content
Merck
All Photos(1)

Documents

116092

Sigma-Aldrich

Triethyl 4-phosphonocrotonate, mixture of isomers

technical grade, 90%

Synonym(s):

4-(Diethoxyphosphinyl)-2-butenoic acid ethyl ester, 4-Phosphonocrotonic acid triethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH2CH=CHCO2C2H5
CAS Number:
Molecular Weight:
250.23
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.455 (lit.)

bp

135 °C/0.4 mmHg (lit.)

density

1.128 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)\C=C\CP(=O)(OCC)OCC

InChI

1S/C10H19O5P/c1-4-13-10(11)8-7-9-16(12,14-5-2)15-6-3/h7-8H,4-6,9H2,1-3H3/b8-7+

InChI key

LXLODBXSCRTXFG-BQYQJAHWSA-N

Application

Reactant for:
  • Iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase
  • Stereoselective preparation of C1-C19 fragment of macrolide antibiotic aplyronine A using diastereoselective Nozaki-Hiyama-Kishi coupling reactions
  • Orally bioavailable GPR40 agonist synthesis
  • Synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2
  • Preaparation of mGlu4R agonists
  • Dearomatizing cyclization of nicotinyl-substituted esters and ketones

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service