Merck
  • Effects of bioisosteric fluorine in synthetic cannabinoid designer drugs JWH-018, AM-2201, UR-144, XLR-11, PB-22, 5F-PB-22, APICA, and STS-135.
Effects of bioisosteric fluorine in synthetic cannabinoid designer drugs JWH-018, AM-2201, UR-144, XLR-11, PB-22, 5F-PB-22, APICA, and STS-135.
ACS chemical neuroscience (2015-04-30)
Samuel D Banister, Jordyn Stuart, Richard C Kevin, Amelia Edington, Mitchell Longworth, Shane M Wilkinson, Corinne Beinat, Alexandra S Buchanan, David E Hibbs, Michelle Glass, Mark Connor, Iain S McGregor, Michael Kassiou
ABSTRACT
Synthetic cannabinoid (SC) designer drugs featuring bioisosteric fluorine substitution are identified by forensic chemists and toxicologists with increasing frequency. Although terminal fluorination of N-pentyl indole SCs is sometimes known to improve cannabinoid type 1 (CB1) receptor binding affinity, little is known of the effects of fluorination on functional activity of SCs. This study explores the in vitro functional activities of SC designer drugs JWH-018, UR-144, PB-22, and APICA, and their respective terminally fluorinated analogues AM-2201, XLR-11, 5F-PB-22, and STS-135 at human CB1 and CB2 receptors using a FLIPR membrane potential assay. All compounds demonstrated agonist activity at CB1 (EC50 = 2.8-1959 nM) and CB2 (EC50 = 6.5-206 nM) receptors, with the fluorinated analogues generally showing increased CB1 receptor potency (∼2-5 times). Additionally, the cannabimimetic activities and relative potencies of JWH-018, AM-2201, UR-144, XLR-11, PB-22, 5F-PB-22, APICA, and STS-135 in vivo were evaluated in rats using biotelemetry. All SCs dose-dependently induced hypothermia and reduced heart rate at doses of 0.3-10 mg/kg. There was no consistent trend for increased potency of fluorinated SCs over the corresponding des-fluoro SCs in vivo. Based on magnitude and duration of hypothermia, the SCs were ranked for potency (PB-22 > 5F-PB-22 = JWH-018 > AM-2201 > APICA = STS-135 = XLR-11 > UR-144).
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%