Skip to Content
Merck
All Photos(1)

Documents

09-1320

Sigma-Aldrich

Ethylenediaminetetraacetic acid

SAJ special grade, ≥99.0%

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
292.24
Beilstein:
1716295
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

powder

availability

available only in Japan

mp

250 °C (dec.) (lit.)

density

1.46 g/cm3 at 20 °C

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucienne Tritten et al.
PLoS neglected tropical diseases, 8(7), e2971-e2971 (2014-07-18)
Filarial nematodes cause chronic and profoundly debilitating diseases in both humans and animals. Applications of novel technology are providing unprecedented opportunities to improve diagnosis and our understanding of the molecular basis for host-parasite interactions. As a first step, we investigated
Cecilia Lezcano et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 27(9), 1193-1202 (2014-01-18)
Of more than 150 000 published studies evaluating new biomarkers, fewer than 100 biomarkers have been implemented for patient care. One reason for this is lack of rigorous testing by the medical community to validate claims for biomarker clinical relevance, and
Derek K Chu et al.
The Journal of experimental medicine, 211(8), 1657-1672 (2014-07-30)
Eosinophils natively inhabit the small intestine, but a functional role for them there has remained elusive. Here, we show that eosinophil-deficient mice were protected from induction of Th2-mediated peanut food allergy and anaphylaxis, and Th2 priming was restored by reconstitution
Khosrow Adeli et al.
Clinical chemistry, 61(8), 1063-1074 (2015-06-06)
Defining laboratory biomarker reference values in a healthy population and understanding the fluctuations in biomarker concentrations throughout life and between sexes are critical to clinical interpretation of laboratory test results in different disease states. The Canadian Health Measures Survey (CHMS)
Huan Shu et al.
Genome biology, 15(4), R62-R62 (2014-04-09)
Histone variants establish structural and functional diversity of chromatin by affecting nucleosome stability and histone-protein interactions. H3.3 is an H3 histone variant that is incorporated into chromatin outside of S-phase in various eukaryotes. In animals, H3.3 is associated with active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service