Merck
  • The effects of under 6 hours of formalin fixation on hormone receptor and HER2 expression in invasive breast cancer: a systematic review.
The effects of under 6 hours of formalin fixation on hormone receptor and HER2 expression in invasive breast cancer: a systematic review.
American journal of clinical pathology (2014-06-14)
Shona Kalkman, Maarten W Barentsz, Paul J van Diest
ABSTRACT
A systematic review of the literature was performed to identify whether minimum formalin fixation time may be reduced for reliable immunohistochemical assessment of estrogen receptor (ER), progesterone receptor (PR), and human epidermal growth factor receptor 2 (HER2). PubMed, EMBASE, and the Cochrane Library were systematically searched for studies addressing effects of brief tissue fixation (<6 hours) on the analysis of ER, PR, or HER2 expression in patients with breast cancer. Five publications reported effects of brief fixation on ER, PR, or HER2 expression. Four studies showed similar receptor expression of short fixation compared with recommended fixation time (6-72 hours). One publication found that a minimum fixation time of 6 to 8 hours is necessary for reliable ER results. Available data on the effect of brief fixation on receptor status are limited. However, brief fixation of very highly expressing breast cancers does not seem to alter ER, PR, and HER2 status. Nevertheless, scoring inconsistencies have been observed. Further research is required in larger study populations with more low-expressing cases for future validation.
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O