Skip to Content
Merck
All Photos(1)

Key Documents

11-0706

Sigma-Aldrich

Formaldehyde neutral buffer solution

7.5% formaldehyde in H2O

Synonym(s):

Formaldehyde solution, Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

availability

available only in Japan

concentration

7.5% formaldehyde in H2O

pH

7.10-7.30 (20 °C)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdel Nasser Hosein et al.
Laboratory investigation; a journal of technical methods and pathology, 93(6), 701-710 (2013-04-10)
Pathology archives contain vast resources of clinical material in the form of formalin-fixed paraffin-embedded (FFPE) tissue samples. Owing to the methods of tissue fixation and storage, the integrity of DNA and RNA available from FFPE tissue is compromized, which means
Anne M Mills et al.
The American journal of surgical pathology, 37(7), 995-1000 (2013-05-08)
Cytomegalovirus (CMV) can precipitate and exacerbate gastrointestinal (GI) mucosal injury. The gold standard for CMV detection in formalin-fixed, paraffin-embedded (FFPE) tissue is immunohistochemistry (IHC). Although CMV polymerase chain reaction (PCR) on fresh tissue may be a valuable adjunct to IHC
M M Kamieniak et al.
British journal of cancer, 108(8), 1732-1742 (2013-04-06)
Few studies have attempted to characterise genomic changes occurring in hereditary epithelial ovarian carcinomas (EOCs) and inconsistent results have been obtained. Given the relevance of DNA copy number alterations in ovarian oncogenesis and growing clinical implications of the BRCA-gene status
Célina Roda et al.
Environmental research, 123, 46-51 (2013-04-09)
Although formaldehyde is a common indoor pollutant, its impact on respiratory symptoms in childhood remains unclear. The aim of this study was to examine the relation between domestic formaldehyde exposure and occurrence of coughing, one of the most prevalent respiratory
R Mezzapelle et al.
British journal of cancer, 108(8), 1743-1749 (2013-04-06)
As epidermal growth factor receptor (EGFR) is involved in the pathogenesis of malignant pleural mesotheliomas (MPMs), the anti-EGFR drugs may be effective in treating MPM patients. Mutations of the EGFR gene or its downstream effectors may cause constitutive activation leading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service