Merck
  • Brief fixation does not affect assessment of hormone receptor expression in invasive breast carcinoma biopsies: paving the road for same-day tissue diagnostics.
Brief fixation does not affect assessment of hormone receptor expression in invasive breast carcinoma biopsies: paving the road for same-day tissue diagnostics.
The American journal of surgical pathology (2014-04-08)
Shona Kalkman, Maarten W Barentsz, Arjen J Witkamp, Elsken van der Wall, Helena M Verkooijen, Paul J van Diest
ABSTRACT
In patients with invasive breast carcinoma, estrogen receptor α (ERα) and progesterone receptor (PR) expressions need to be assessed in core-needle biopsies (CNBs) before the start of neoadjuvant systemic treatment. Current guidelines recommend a minimum formalin fixation time of 6 hours. Considering the increasing demand for same-day diagnostics in oncology, more rapid tissue processing with shorter fixation times is required. To identify whether brief fixation (<6 h) of CNBs compared with conventionally fixed resection specimens provides for reliable immunohistochemical assessment of ERα and PR expression, 78 consecutive patients diagnosed with invasive breast carcinoma were included through the same-day diagnostics programme of the UMC Utrecht. Paraffin-embedded CNBs fixed for approximately 45 minutes were retrieved. Immunohistochemistry for ERα and PR was compared between the briefly fixed CNBs and conventionally fixed resection specimens. All slides were reviewed by means of consensus scoring by 2 blinded observers. Overall agreement between CNB and resections was 73/74 (98.6%) for ERα (κ=0.85; 95% confidence interval [CI]=0.56-1.00) and 69/75 (92.0%) for PR (κ=0.81; 95% CI=0.66-0.96). For ERα, positive and negative predictive values were 98.6% (95% CI=0.91-0.99) and 100.0% (95% CI=0.31-1.00), respectively. For PR, positive and negative predictive values were 100.0% (95% CI=0.91-1.00) and 76.0% (95% CI=0.54-0.90). In conclusion, analysis of hormone receptor expression in briefly fixed CNB seems comparable to results from conventionally fixed resection specimens of the same tumor.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O