PHR9270
Cystamine dihydrochloride
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
Agency
(Not traceable to any primary pharmacopeia standard)
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
mp
217-220 °C (dec.) (lit.)
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
Cl[H].Cl[H].NCCSSCCN
InChI
1S/C4H12N2S2.2ClH/c5-1-3-7-8-4-2-6;;/h1-6H2;2*1H
InChI key
YUFRRMZSSPQMOS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service