Skip to Content
Merck
All Photos(1)

Key Documents

842177

TSKgel® Super-Phenyl (2.3 µm) HPLC Columns

L x I.D. 1 cm × 2 mm, phenyl phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® Super-Phenyl Guard Cartridge, phase phenyl, L × I.D. 1 cm × 2 mm, 2.3 μm particle size

material

stainless steel

Agency

suitable for USP L11

product line

TSKgel®

manufacturer/tradename

Tosoh 42177

extent of labeling

3% Carbon, multilayer, endcapped

parameter

no limitation salt concentration
0-100% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1 cm × 2 mm

surface area

100 m2/g

matrix

spherical silica

matrix active group

phenyl phase

particle size

2.3 μm

operating pH

2.0-7.5

separation technique

reversed phase

General description

Requires Guard Cartridge Holder, PN 819308.
TSKgel® Super-Phenyl columns are packed with monodispersed 2 micron (nominal, mean particle size is 2.3 micron) spherical silica particles covalently bonded with phenyl groups. The small particle size used in Super Series and TSKgel ODS-140HTP columns provides the highest efficiency of all TSKgel reversed phase columns. The monodispersity of the particles reduces column back pressure to values more typical of larger particles, thus allowing the use of higher flow rates than other 2 micron packings. TSKgel Super-Phenyl columns are less hydrophobic than TSKgel Super-Octyl and TSKgel Super-ODS columns. They are an excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules and food/beverage samples.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

accessory

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service