Skip to Content
Merck
All Photos(1)

Key Documents

57665-U

Upchurch Polymer Tubing Cutter

Synonym(s):

Polymer Tubing Cutting Tool

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.

usage

for cutting 1/8-1/16" polymer tubing

General description

Upchurch designed this reliable and durable device specifically for cutting 1/8" - 1/16" PEEK, PTFE, and Tefzel tubing. Includes 4 replacement blades.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. Can I order extra blades for this cutter? If so what is the part number?

    1 answer
    1. Yes, please see product number 57666-U: https://www.sigmaaldrich.com/product/supelco/57666-u.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service